Gene Transcripts

Gene Symbol
PABPC1
Entrez Gene ID
26986
Gene Name
poly(A) binding protein cytoplasmic 1
Gene Aliases
PAB1, PABP, PABP1, PABPC2, PABPL1
Chromosome Location
Chr. 8: 100702916 - 100722998 on Build GRCh38
UniGene ID
Hs.387804
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002568.3 NP_002559.2 15 2682
XM_005250861.2 XP_005250918.1 15 3610
GenBank mRNA AL135187.1 - 1 245
BC015958.2 - 16 2577 3940309
BC041863.1 - 15 3304
Y00345.1 - 15 2670

View Full Product Details