Gene Transcripts

Gene Symbol
KRTAP10-3
Entrez Gene ID
386682
Gene Name
keratin associated protein 10-3
Gene Aliases
KAP10.3, KAP18-3, KAP18.3, KRTAP10.3, KRTAP18-3, KRTAP18.3
Chromosome Location
Chr. 21: 44558023 - 44558760 on Build GRCh38
UniGene ID
Hs.688630
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198696.2 NP_941969.2 1 658
GenBank mRNA AB076350.1 - 1 658
AJ566383.1 - 1 117
BC133677.1 - 2 556 40119288

View Full Product Details