Gene Transcripts

Gene Symbol
MRVI1
Entrez Gene ID
10335
Gene Name
murine retrovirus integration site 1 homolog
Gene Aliases
IRAG, JAW1L
Chromosome Location
Chr. 11: 10573091 - 10693988 on Build GRCh38
UniGene ID
Hs.501898
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098579.2 NP_001092049.2 3 410
NM_001100163.2 NP_001093633.1 4 691
NM_001100167.2 NP_001093637.1 4 405
NM_001206880.1 NP_001193809.1 4 744
NM_001206881.1 NP_001193810.1 3 586
NM_130385.3 NP_569056.4 4 744
GenBank mRNA AF081249.1 - 3 483
AF081250.1 - 3 211
AK127209.1 - 4 383
AK296336.1 - 4 508
AK300358.1 - 3 395
AK301296.1 - 4 355
AK304702.1 - 4 378
BC117127.1 - 4 281 40125678
DA127310.1 - 3 563
DA797538.1 - 3 410
DB132030.1 - 3 203
DB167290.1 - 5 493
DB445355.2 - 5 627

View Full Product Details