Gene Transcripts
- Gene Symbol
- TTC31
- Entrez Gene ID
- 64427
- Gene Name
- tetratricopeptide repeat domain 31
- Gene Aliases
- Chromosome Location
- Chr. 2: 74483032 - 74494564 on Build GRCh38
- UniGene ID
- Hs.557709
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022492.4 |
NP_071937.4
|
2, 3 |
160 |
|
XM_005264492.1 |
XP_005264549.1
|
2, 3 |
149 |
|
XM_011533035.1 |
XP_011531337.1
|
2, 3 |
149 |
|
XM_011533036.1 |
XP_011531338.1
|
2, 3 |
149 |
|
XM_011533037.1 |
XP_011531339.1
|
2, 3 |
149 |
|
XM_017004706.1 |
XP_016860195.1
|
2, 3 |
149 |
|
XM_017004707.1 |
XP_016860196.1
|
2, 3 |
149 |
|
GenBank mRNA |
AK022850.1 |
- |
2, 3 |
146 |
|
AK223355.1 |
- |
2, 3 |
144 |
|
AK225626.1 |
- |
2, 3 |
146 |
|
AK301585.1 |
- |
2, 3 |
144 |
|
AK303245.1 |
- |
2, 3 |
146 |
|
BC036017.1 |
- |
2, 3 |
144 |
5270575 |
BC096709.1 |
- |
2, 3 |
146 |
40023059 |
BC098365.1 |
- |
2, 3 |
146 |
40023060 |
View Full Product Details