Gene Transcripts

Gene Symbol
SLC25A18
Entrez Gene ID
83733
Gene Name
solute carrier family 25 member 18
Gene Aliases
GC2
Chromosome Location
Chr. 22: 17562470 - 17590890 on Build GRCh38
UniGene ID
Hs.282982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303484.1 NP_001290413.1 10 1813
NM_031481.2 NP_113669.1 11 1876
XM_011546149.2 XP_011544451.1 11 2805
XM_011546150.2 XP_011544452.1 11 2675
XM_011546151.2 XP_011544453.1 11 2064
XM_011546152.1 XP_011544454.1 11 1691
XM_011546153.2 XP_011544455.1 11 2686
XM_011546154.2 XP_011544456.1 11 1933
XM_017028968.1 XP_016884457.1 11 1559
XM_017028969.1 XP_016884458.1 11 3705
XM_017028970.1 XP_016884459.1 12 3196
XM_017028971.1 XP_016884460.1 12 3185
XM_017028972.1 XP_016884461.1 12 3108
XM_017028973.1 XP_016884462.1 11 3133
XM_017028974.1 XP_016884463.1 12 3064
XM_017028975.1 XP_016884464.1 12 2976
GenBank mRNA AJ428203.1 - 11 1832
AY008285.1 - 11 1832
BC031644.1 - 10 1768 5179038

View Full Product Details