Gene Transcripts

Gene Symbol
REEP6
Entrez Gene ID
92840
Gene Name
receptor accessory protein 6
Gene Aliases
C19orf32, DP1L1, TB2L1, Yip2f
Chromosome Location
Chr. 19: 1491166 - 1497925 on Build GRCh38
UniGene ID
Hs.76277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138393.1 NP_612402.1 5 1356
GenBank mRNA AK058112.1 - 6 1331
AX775999.1 - 5 1356
BC008201.1 - 5 1356 3865853

View Full Product Details