Gene Transcripts

Gene Symbol
OCIAD2
Entrez Gene ID
132299
Gene Name
OCIA domain containing 2
Gene Aliases
Chromosome Location
Chr. 4: 48885380 - 48906828 on Build GRCh38
UniGene ID
Hs.95835
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014446.2 NP_001014446.1 4, 5 458
NM_001286773.1 NP_001273702.1 4, 5 481
NM_001286774.1 NP_001273703.1 3, 4 361
NM_152398.3 NP_689611.1 4, 5 458
GenBank mRNA AK298305.1 - 4, 5 427
AY472117.1 - 4, 5 460
BC032808.1 - 4, 5 429 4454708
BG748993.1 - 4, 5 364
BI546814.1 - 4, 5 463
BM782144.1 - 4, 5 428
BU508058.1 - 3, 4 249
CR749310.1 - 4, 5 403
W68799.1 - 4, 5 410

View Full Product Details