Gene Transcripts

Gene Symbol
RAD54B
Entrez Gene ID
25788
Gene Name
RAD54 homolog B (S. cerevisiae)
Gene Aliases
RDH54
Chromosome Location
Chr. 8: 94371960 - 94475115 on Build GRCh38
UniGene ID
Hs.30561
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205263.1 NP_001192192.1 13 2818
NM_012415.3 NP_036547.1 15 2795
GenBank mRNA AF086020.1 - 1 19
AF112481.1 - 15 2744
AK290437.1 - 15 2765
AL133578.1 - 9 1676
BC001965.2 - 15 2737 3452509

View Full Product Details