Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243998.1 | NP_001230927.1 | 4 | 208 | |
NM_001243999.1 | NP_001230928.1 | 5 | 423 | ||
NM_001244000.1 | NP_001230929.1 | 5 | 388 | ||
NM_003121.4 | NP_003112.2 | 5 | 423 | ||
GenBank mRNA | AK225225.1 | - | 5 | 421 | |
AK225720.1 | - | 5 | 423 | ||
AK292644.1 | - | 5 | 425 | ||
AK300639.1 | - | 4 | 206 | ||
BC007921.2 | - | 5 | 402 | 4309499 | |
DA957263.1 | - | 5 | 385 | ||
X66079.1 | - | 5 | 403 | ||
X96998.1 | - | 5 | 403 |