Gene Transcripts
- Gene Symbol
- Tfpi2
- Entrez Gene ID
- 286926
- Gene Name
- tissue factor pathway inhibitor 2
- Gene Aliases
- Chromosome Location
- Chr. 4: 28948343 - 28952765 on Build Rnor_6.0
- UniGene ID
- Rn.15776
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173141.1 |
NP_775164.1
|
1, 2 |
148 |
|
GenBank mRNA |
AJ428954.1 |
- |
1, 2 |
148 |
|
View Full Product Details