Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001109975.1 | NP_001103445.1 | 2 | 2056 | |
NM_177340.2 | NP_796314.2 | 2 | 1997 | ||
GenBank mRNA | AK129267.1 | - | 2 | 1947 | |
AK141564.1 | - | 2 | 2058 | ||
AK154958.1 | - | 2 | 1992 | ||
AK158933.1 | - | 3 | 2020 | ||
AK172163.1 | - | 2 | 1951 | ||
BC065127.1 | - | 2 | 2263 |