Gene Transcripts

Gene Symbol
Sult2a1
Entrez Gene ID
20859
Gene Name
sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 1
Gene Aliases
ST2A1, Std, Sth1, mSTa1
Chromosome Location
Chr. 7: 13796246 - 13837410 on Build GRCm38
UniGene ID
Mm.260026
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111296.2 NP_001104766.1 4 592
GenBank mRNA BC145167.1 - 4 564
L02335.1 - 4 565

View Full Product Details