Gene Transcripts
- Gene Symbol
- BTAF1
- Entrez Gene ID
- 9044
- Gene Name
- B-TFIID TATA-box binding protein associated factor 1
- Gene Aliases
- MOT1, TAF(II)170, TAF172, TAFII170
- Chromosome Location
- Chr. 10: 91923769 - 92030998 on Build GRCh38
- UniGene ID
- Hs.500526
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003972.2 |
NP_003963.1
|
12, 13 |
1510 |
|
XM_011540327.2 |
XP_011538629.1
|
9, 10 |
1275 |
|
XM_011540328.2 |
XP_011538630.1
|
9, 10 |
1103 |
|
XM_011540329.2 |
XP_011538631.1
|
12, 13 |
1720 |
|
XM_017016877.1 |
XP_016872366.1
|
11, 12 |
1572 |
|
XM_017016878.1 |
XP_016872367.1
|
11, 12 |
1402 |
|
XM_017016879.1 |
XP_016872368.1
|
11, 12 |
1526 |
|
XM_017016880.1 |
XP_016872369.1
|
11, 12 |
1530 |
|
XM_017016881.1 |
XP_016872370.1
|
10, 11 |
1379 |
|
GenBank mRNA |
AF038362.1 |
- |
12, 13 |
1529 |
|
AJ001017.2 |
- |
12, 13 |
1424 |
|
BC030273.1 |
- |
6, 7 |
757 |
|
BC112201.1 |
- |
12, 13 |
1494 |
8327669 |
View Full Product Details