Gene Transcripts

Gene Symbol
Slc30a2
Entrez Gene ID
230810
Gene Name
solute carrier family 30 (zinc transporter), member 2
Gene Aliases
6030430C21, ZnT-2, Znt2
Chromosome Location
Chr. 4: 134342961 - 134356528 on Build GRCm38
UniGene ID
Mm.358876
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039677.2 NP_001034766.1 4 721
GenBank mRNA AK031425.1 - 4 517
BC113751.1 - 4 721 40057211

View Full Product Details