Gene Transcripts

Gene Symbol
Ssxb6
Entrez Gene ID
668976
Gene Name
synovial sarcoma, X member B, breakpoint 6
Gene Aliases
Chromosome Location
Chr. X: 8542179 - 8548362 on Build GRCm38
UniGene ID
Mm.227643
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205108.1 NP_001192037.1 150
GenBank mRNA AY244797.1 - 151

View Full Product Details