Gene Transcripts
- Gene Symbol
- Rps10
- Entrez Gene ID
- 81773
- Gene Name
- ribosomal protein S10
- Gene Aliases
- Chromosome Location
- Chr. 20: 7215762 - 7220331 on Build Rnor_6.0
- UniGene ID
- Rn.3359
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031109.1 |
NP_112371.1
|
2, 3 |
196 |
|
GenBank mRNA |
BC058141.1 |
- |
2, 3 |
184 |
|
FQ210550.1 |
- |
2, 3 |
187 |
|
FQ217175.1 |
- |
2, 3 |
185 |
|
FQ217183.1 |
- |
2, 3 |
186 |
|
FQ221004.1 |
- |
2, 3 |
186 |
|
FQ221394.1 |
- |
2, 3 |
186 |
|
FQ221397.1 |
- |
2, 3 |
186 |
|
FQ221701.1 |
- |
2, 3 |
186 |
|
FQ221937.1 |
- |
2, 3 |
186 |
|
FQ222456.1 |
- |
2, 3 |
191 |
|
FQ222595.1 |
- |
2, 3 |
189 |
|
FQ222615.1 |
- |
2, 3 |
188 |
|
FQ222830.1 |
- |
2, 3 |
186 |
|
FQ223309.1 |
- |
2, 3 |
188 |
|
FQ223371.1 |
- |
2, 3 |
177 |
|
FQ224021.1 |
- |
2, 3 |
186 |
|
FQ228540.1 |
- |
2, 3 |
186 |
|
X13549.1 |
- |
2, 3 |
196 |
|
- Gene Symbol
- Rpl30
- Entrez Gene ID
- 64640
- Gene Name
- ribosomal protein L30
- Gene Aliases
- Chromosome Location
- Chr. 7: 73213538 - 73216431 on Build Rnor_6.0
- UniGene ID
- Rn.161950
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
FQ229603.1 |
- |
|
186 |
|
- Gene Symbol
- Rps10l1
- Entrez Gene ID
- 100363439
- Gene Name
- ribosomal protein S10-like 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 75429715 - 75430282 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details