Gene Transcripts
- Gene Symbol
- NOLC1
- Entrez Gene ID
- 9221
- Gene Name
- nucleolar and coiled-body phosphoprotein 1
- Gene Aliases
- NOPP130, NOPP140, NS5ATP13, P130
- Chromosome Location
- Chr. 10: 102152176 - 102163871 on Build GRCh38
- UniGene ID
- Hs.523238
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001284388.1 |
NP_001271317.1
|
11, 12 |
2114 |
|
NM_001284389.1 |
NP_001271318.1
|
11, 12 |
2087 |
|
NM_004741.4 |
NP_004732.2
|
11, 12 |
2084 |
|
XM_005270273.1 |
XP_005270330.1
|
11, 12 |
2091 |
|
GenBank mRNA |
AK056755.1 |
- |
10, 11 |
1707 |
|
AK314363.1 |
- |
11, 12 |
1871 |
|
AY820769.1 |
- |
11, 12 |
1852 |
|
BC001883.1 |
- |
11, 12 |
1859 |
3533568 |
BC006769.2 |
- |
7, 8 |
1369 |
2900024 |
CN312242.1 |
- |
2, 3 |
270 |
|
D21262.1 |
- |
11, 12 |
1875 |
|
Z34289.1 |
- |
11, 12 |
1903 |
|
View Full Product Details