Gene Transcripts
- Gene Symbol
- Rbm45
- Entrez Gene ID
- 266631
- Gene Name
- RNA binding motif protein 45
- Gene Aliases
- Drb1
- Chromosome Location
- Chr. 3: 63220660 - 63235638 on Build Rnor_6.0
- UniGene ID
- Rn.204106
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153306.1 |
NP_695218.1
|
1, 2 |
358 |
|
GenBank mRNA |
AB036990.1 |
- |
1, 2 |
358 |
|
View Full Product Details