Gene Transcripts

Gene Symbol
RXFP1
Entrez Gene ID
59350
Gene Name
relaxin/insulin like family peptide receptor 1
Gene Aliases
LGR7, RXFPR1
Chromosome Location
Chr. 4: 158521711 - 158653372 on Build GRCh38
UniGene ID
Hs.591686
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253727.1 NP_001240656.1 2 340
NM_001253728.1 NP_001240657.1 2 340
NM_001253729.1 NP_001240658.1 2 340
NM_001253730.1 NP_001240659.1 2 340
NM_001253732.1 NP_001240661.1 2 340
NM_001253733.1 NP_001240662.1 2 340
NM_021634.3 NP_067647.2 2 340
NR_045579.1 3 1029
NR_045580.1 2 340
NR_045581.1 2 340
NR_045582.1 2 340
NR_045584.1 2 340
XM_011532174.1 XP_011530476.1 2 309
XM_011532175.1 XP_011530477.1 2 309
XM_011532176.1 XP_011530478.1 2 312
XM_011532179.2 XP_011530481.1 2 309
XM_017008517.1 XP_016864006.1 2 309
XM_017008518.1 XP_016864007.1 2 312
XM_017008519.1 XP_016864008.1 3 273
XM_017008520.1 XP_016864009.1 2 338
XM_017008521.1 XP_016864010.1 3 273
XM_017008522.1 XP_016864011.1 2 337
XM_017008523.1 XP_016864012.1 2 312
XM_017008524.1 XP_016864013.1 2 312
XM_017008525.1 XP_016864014.1 2 343
GenBank mRNA AB208943.1 - 2 78
AF190500.1 - 2 77
AK122647.1 - 4 381
AK294694.1 - 2 104
AK295040.1 - 3 852
AK300379.1 - 2 235
AK312470.1 - 2 172
AY899848.1 - 2 77
AY899849.1 - 2 77
AY899850.1 - 2 77
BC112142.1 - 2 159 8327610
BC113617.1 - 2 162 8322669
BC143556.1 - 2 158
BC143558.1 - 2 162
BC143559.1 - 2 162
BX647975.1 - 2 278
BX647980.1 - 2 278
BX647985.1 - 2 307
DA241507.1 - 2 313
DA519897.1 - 2 203
DC334052.1 - 2 342

View Full Product Details