Gene Transcripts

Gene Symbol
Sycn
Entrez Gene ID
245917
Gene Name
syncollin
Gene Aliases
Syl
Chromosome Location
Chr. 1: 85219827 - 85220239 on Build Rnor_6.0
UniGene ID
Rn.11220
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139086.1 NP_620786.1 1 123
GenBank mRNA AF008197.1 - 1 123
AF012887.1 - 103

View Full Product Details