Gene Transcripts

Gene Symbol
Reep5
Entrez Gene ID
13476
Gene Name
receptor accessory protein 5
Gene Aliases
AI324241, AU022809, AW495741, DP1/TB2, Dp1, R74856, TB2/DP1
Chromosome Location
Chr. 18: 34344885 - 34373415 on Build GRCm38
UniGene ID
Mm.280075
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007874.3 NP_031900.3 5 1899
GenBank mRNA AK013146.1 - 5 1830
BC013052.1 - 5 1830 3585290

View Full Product Details