Gene Transcripts

Gene Symbol
Gpr3
Entrez Gene ID
14748
Gene Name
G-protein coupled receptor 3
Gene Aliases
Gpcr20, Gpcr21, Gpcr3
Chromosome Location
Chr. 4: 133209340 - 133212536 on Build GRCm38
UniGene ID
Mm.4721
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008154.3 NP_032180.1 2 1935
GenBank mRNA AK140302.1 - 2 1906
AK140464.1 - 2 1889
AK158917.1 - 2 1898
D21062.1 - 1 2310

View Full Product Details