Gene Transcripts

Gene Symbol
Ppt1
Entrez Gene ID
19063
Gene Name
palmitoyl-protein thioesterase 1
Gene Aliases
9530043G02Rik, AA960502, C77813, CLN1, D4Ertd184e, INCL, PPT
Chromosome Location
Chr. 4: 122836075 - 122859175 on Build GRCm38
UniGene ID
Mm.277719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008917.3 NP_032943.2 3 358
GenBank mRNA AF071025.1 - 3 375
AF326557.1 - 3 342
AF326558.1 - 3 342
AK014561.1 - 3 340
AK148314.1 - 3 333
AK151055.1 - 3 333
AK151107.1 - 3 344
AK152474.1 - 3 342
AK152940.1 - 3 344
AK153106.1 - 3 339
AK153316.1 - 3 339
AK169579.1 - 4 433
AK169658.1 - 4 439
AK171668.1 - 3 340
AK192077.1 - 1 13
AK192572.1 - 1 13
AK195785.1 - 1 13
CB600397.1 - 3 361

View Full Product Details