Gene Transcripts

Gene Symbol
Vpreb3
Entrez Gene ID
22364
Gene Name
pre-B lymphocyte gene 3
Gene Aliases
8HS-20, AI528709, Vpreb-3
Chromosome Location
Chr. 10: 75943025 - 75949651 on Build GRCm38
UniGene ID
Mm.333851
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009514.4 NP_033540.1 2 378
GenBank mRNA AK008794.1 - 2 379
BC062250.1 - 2 358 1246296

View Full Product Details