Gene Transcripts

Gene Symbol
Sbpl
Entrez Gene ID
638345
Gene Name
spermine binding protein-like
Gene Aliases
2310068J22Rik
Chromosome Location
Chr. 17: 23953085 - 23955273 on Build GRCm38
UniGene ID
Mm.46428
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077421.1 NP_001070889.1 2 190
GenBank mRNA AK009270.1 - 2 192
AK009356.1 - 2 180
AK009376.1 - 2 191
AK009398.1 - 2 192
AK009406.1 - 2 191
AK009446.1 - 2 191
AK009467.1 - 2 191
AK009489.1 - 2 191
AK009501.1 - 2 189
AK009509.1 - 2 191
AK009539.1 - 2 191
AK009553.1 - 2 190
AK009598.1 - 2 191
AK009609.1 - 2 191
AK009613.1 - 2 191
AK009627.1 - 2 193
AK009630.1 - 2 190
AK009631.1 - 2 194
AK009647.1 - 2 189
AK009712.1 - 2 191
AK009758.1 - 2 189
AK009761.1 - 2 189
AK009792.1 - 2 189
AK009819.1 - 2 191
AK009834.1 - 2 191
AK009845.1 - 2 191
AK009910.1 - 2 190
AK009917.1 - 2 192
AK009931.1 - 2 193
AK009982.1 - 2 190
AK010076.1 - 2 190
AK010078.1 - 2 191
AK010083.1 - 2 191
AK010102.1 - 2 191
AK010114.1 - 2 191
AK010161.1 - 2 191
AK010189.1 - 2 191
AK010216.1 - 2 191
AK010221.1 - 2 190
AK010237.1 - 2 190
AK075878.1 - 1 70
AK160518.1 - 2 190
BC089471.1 - 2 188 30301294

Gene Symbol
Sbp
Entrez Gene ID
20234
Gene Name
spermine binding protein
Gene Aliases
AV082137, AV082259, p25
Chromosome Location
Chr. 17: 23941996 - 23945607 on Build GRCm38
UniGene ID
Mm.390958
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011321.3 NP_035451.1 2 205
GenBank mRNA AK035423.1 - 2 206
BC049605.1 - 2 205 6743567
X06246.1 - 2 205

View Full Product Details