Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011403.2 | NP_035533.1 | 20 | 3208 | |
GenBank mRNA | AK137542.1 | - | 20 | 3104 | |
AK157181.1 | - | 20 | 3177 | ||
AK157833.1 | - | 20 | 3066 | ||
AK158243.1 | - | 20 | 3078 | ||
AK158453.1 | - | 20 | 3053 | ||
AK162616.1 | - | 20 | 3053 | ||
AK165195.1 | - | 20 | 3065 | ||
AK165267.1 | - | 20 | 3072 | ||
AK171808.1 | - | 20 | 3042 | ||
AK172103.1 | - | 20 | 3065 | ||
AK172316.1 | - | 20 | 3053 | ||
AK172395.1 | - | 20 | 3104 | ||
AK172456.1 | - | 20 | 3088 | ||
AK172569.1 | - | 20 | 3053 | ||
AY296129.1 | - | 20 | 3042 | ||
BC052419.1 | - | 20 | 3048 | 5702599 | |
BC053429.1 | - | 20 | 3039 | 6506270 | |
M29379.1 | - | 20 | 3036 | ||
X02677.1 | - | 20 | 3042 | ||
X03917.1 | - | 18 | 2885 |