Gene Transcripts

Gene Symbol
Zim1
Entrez Gene ID
22776
Gene Name
zinc finger, imprinted 1
Gene Aliases
Chromosome Location
Chr. 7: 6674444 - 6696538 on Build GRCm38
UniGene ID
Mm.89976
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011769.4 NP_035899.4 5, 6 430
GenBank mRNA AF111101.1 - 6, 7 553
AK086590.1 - 5, 6 449
BC066196.1 - 5, 6 430 30430385

View Full Product Details