Gene Transcripts
- Gene Symbol
- Rnf13
- Entrez Gene ID
- 24017
- Gene Name
- ring finger protein 13
- Gene Aliases
- 2010001H16Rik, Rzf
- Chromosome Location
- Chr. 3: 57736043 - 57835233 on Build GRCm38
- UniGene ID
- Mm.274360
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF037205.1 |
- |
3, 4 |
283 |
|
AF037206.1 |
- |
3, 4 |
283 |
|
AI882163.1 |
- |
3, 4 |
342 |
|
AK034135.1 |
- |
3, 4 |
334 |
|
AK036591.1 |
- |
3, 4 |
328 |
|
AK050095.1 |
- |
3, 4 |
296 |
|
AK082296.1 |
- |
3, 4 |
324 |
|
AK139447.1 |
- |
3, 4 |
289 |
|
AK140682.1 |
- |
3, 4 |
338 |
|
AK158046.1 |
- |
3, 4 |
324 |
|
AK167278.1 |
- |
3, 4 |
288 |
|
BB847350.1 |
- |
3, 4 |
349 |
|
BC058182.1 |
- |
3, 4 |
291 |
5324040 |
View Full Product Details