Gene Transcripts

Gene Symbol
Mtf2
Entrez Gene ID
17765
Gene Name
metal response element binding transcription factor 2
Gene Aliases
9230112N11Rik, AA537621, C76717, M96, Pcl2
Chromosome Location
Chr. 5: 108065669 - 108109219 on Build GRCm38
UniGene ID
Mm.257149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253877.1 NP_001240806.1 5, 6 1039
NM_001253878.1 NP_001240807.1 5, 6 758
NM_001253879.1 NP_001240808.1 4, 5 681
NM_001253880.1 NP_001240809.1 2, 3 396
NM_013827.3 NP_038855.2 4, 5 677
GenBank mRNA AB221622.1 - 1, 2 84
AK030752.1 - 4, 5 627
AK031409.1 - 6, 7 746
AK032798.1 - 5, 6 1000
AK032819.1 - 5, 6 1001
AK137539.1 - 6, 7 986
AK139685.1 - 5, 6 759
BC024889.1 - 4, 5 649
BC029076.1 - 4, 5 588
BC050826.1 - 4, 5 631
BC057163.1 - 4, 5 474
BC071223.1 - 2, 3 149
BC092237.1 - 4, 5 609 30637248
BC100340.1 - 2, 3 327 30715034
S78454.1 - 1, 2 84

View Full Product Details