Gene Transcripts

Gene Symbol
Mtf2
Entrez Gene ID
17765
Gene Name
metal response element binding transcription factor 2
Gene Aliases
9230112N11Rik, AA537621, C76717, M96, Pcl2
Chromosome Location
Chr. 5: 108065669 - 108109219 on Build GRCm38
UniGene ID
Mm.257149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253877.1 NP_001240806.1 7 1162
NM_001253878.1 NP_001240807.1 7 881
NM_001253879.1 NP_001240808.1 6 804
NM_001253880.1 NP_001240809.1 4 519
NM_013827.3 NP_038855.2 6 800
GenBank mRNA AB221622.1 - 3 207
AK030752.1 - 6 750
AK031409.1 - 8 869
AK032798.1 - 7 1123
AK032819.1 - 7 1124
AK137539.1 - 8 1109
AK139685.1 - 7 882
BC024889.1 - 6 772
BC029076.1 - 6 711
BC050826.1 - 6 754
BC057163.1 - 6 597
BC071223.1 - 4 272
BC092237.1 - 6 732 30637248
BC100340.1 - 4 450 30715034
S78454.1 - 3 207

View Full Product Details