Gene Transcripts

Gene Symbol
Ube4b
Entrez Gene ID
63958
Gene Name
ubiquitination factor E4B
Gene Aliases
4930551I19Rik, 4933406G05Rik, AU014668, D4Bwg0973e, UFD2, UFD2a, Ufd2p, mKIAA0684
Chromosome Location
Chr. 4: 149328416 - 149426637 on Build GRCm38
UniGene ID
Mm.415260
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022022.3 NP_071305.2 4, 5 1169
GenBank mRNA AF260926.1 - 4, 5 1284
AK016689.1 - 4, 5 1170
AK029914.1 - 4, 5 774
AK083409.1 - 4, 5 770
AK122345.1 - 4, 5 680
BC067402.1 - 4, 5 916 30099547
BC075620.1 - 4, 5 1130 6405514

View Full Product Details