Gene Transcripts

Gene Symbol
Stt3b
Entrez Gene ID
68292
Gene Name
STT3, subunit of the oligosaccharyltransferase complex, homolog B (S. cerevisiae)
Gene Aliases
1300006C19Rik, Simp
Chromosome Location
Chr. 9: 115242581 - 115310421 on Build GRCm38
UniGene ID
Mm.296158
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024222.2 NP_077184.2 12 2214
GenBank mRNA AK012153.1 - 12 1884
AK018758.2 - 12 1924
AK145674.1 - 12 1855
AK148065.1 - 12 1913
AK152899.1 - 12 2262
AK154979.1 - 12 1903
AK170079.1 - 12 1926
AK205163.1 - 1 61
AX799084.1 - 12 1925
BC003206.1 - 7 926
BC013054.1 - 2 332
BC052433.1 - 12 2214 6837097

View Full Product Details