Gene Transcripts

Gene Symbol
Slc25a30
Entrez Gene ID
67554
Gene Name
solute carrier family 25, member 30
Gene Aliases
4933433D23Rik, AV025504, AV221431, AW319655, KMCP1
Chromosome Location
Chr. 14: 75757536 - 75787047 on Build GRCm38
UniGene ID
Mm.46067
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026232.3 NP_080508.1 3 297
GenBank mRNA AK015679.1 - 3 298
AK017035.1 - 3 298
AK030950.1 - 3 266
AK090086.1 - 3 235
AK151334.1 - 3 266
AK151420.1 - 3 234
AK159122.1 - 3 295
AK159732.1 - 3 295
BC022676.1 - 3 237 4238839

View Full Product Details