Gene Transcripts

Gene Symbol
Vmn1r4
Entrez Gene ID
171194
Gene Name
vomeronasal 1 receptor 4
Gene Aliases
C230065D10Rik, V1rc21
Chromosome Location
Chr. 6: 56924058 - 56957608 on Build GRCm38
UniGene ID
Mm.222754
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134176.1 NP_598937.1 1 195
GenBank mRNA AK082576.1 - 5 845
BC132386.1 - 1 242 40130663
BC138489.1 - 1 242 8861509

View Full Product Details