Gene Transcripts

Gene Symbol
Vps18
Entrez Gene ID
228545
Gene Name
vacuolar protein sorting 18 (yeast)
Gene Aliases
9930024E13Rik
Chromosome Location
Chr. 2: 119288742 - 119298453 on Build GRCm38
UniGene ID
Mm.294761
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172269.3 NP_758473.3 4 666
GenBank mRNA AK029109.1 - 4 643
AK033333.1 - 4 651
AK036915.1 - 4 651
AK039848.1 - 4 651
AK042471.1 - 4 654
AK044821.1 - 4 669
AK148077.1 - 4 655
AK160689.1 - 4 643
BC036129.1 - 4 594 4972693
BC039176.1 - 4 573 4221232

View Full Product Details