Gene Transcripts

Gene Symbol
Zdhhc9
Entrez Gene ID
208884
Gene Name
zinc finger, DHHC domain containing 9
Gene Aliases
6430508G22, 9530098M12Rik
Chromosome Location
Chr. X: 48171967 - 48208702 on Build GRCm38
UniGene ID
Mm.207367
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172465.4 NP_766053.1 2 459
GenBank mRNA AK032233.1 - 2 441
AK032252.1 - 1 1946
AK035745.1 - 1 1882
AK043810.1 - 2 349
AK079248.1 - 2 426
BC042618.1 - 2 318 4950217
BC090832.1 - 2 277 6401763

View Full Product Details