Gene Transcripts

Gene Symbol
Samd10
Entrez Gene ID
229011
Gene Name
sterile alpha motif domain containing 10
Gene Aliases
D930033N23Rik, E130318E08
Chromosome Location
Chr. 2: 181595218 - 181599491 on Build GRCm38
UniGene ID
Mm.205422
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172676.2 NP_766264.2 5 1708
GenBank mRNA AK053048.1 - 5 1661
AK053884.1 - 3 1289
AK164204.1 - 5 1748
AK171151.1 - 5 1714
AK218993.1 - 1 352
BC052921.1 - 5 1708 6311715

View Full Product Details