Gene Transcripts

Gene Symbol
Tnfrsf26
Entrez Gene ID
244237
Gene Name
tumor necrosis factor receptor superfamily, member 26
Gene Aliases
Tnfrh3, Tnfrsf24
Chromosome Location
Chr. 7: 143607685 - 143627845 on Build GRCm38
UniGene ID
Mm.247498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175649.5 NP_783580.1 8 1133
GenBank mRNA AK142511.1 - 1193
AK154666.1 - 8 1244
AY165628.1 - 8 1133
BC138819.1 - 8 1182 8861841
BC138820.1 - 8 1183 8861842

View Full Product Details