Gene Transcripts
- Gene Symbol
- GYPA
- Entrez Gene ID
- 2993
- Gene Name
- glycophorin A (MNS blood group)
- Gene Aliases
- CD235a, GPA, GPErik, GPSAT, HGpMiV, HGpMiXI, HGpSta(C), MN, MNS, PAS-2
- Chromosome Location
- Chr. 4: 144109303 - 144140854 on Build GRCh38
- UniGene ID
- Hs.434973
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308190.1 |
NP_001295119.1
|
3, 4 |
315 |
|
NM_002099.7 |
NP_002090.4
|
4, 5 |
414 |
|
XM_017008134.1 |
XP_016863623.1
|
4, 5 |
418 |
|
XM_017008135.1 |
XP_016863624.1
|
5, 6 |
481 |
|
XM_017008136.1 |
XP_016863625.1
|
3, 4 |
383 |
|
GenBank mRNA |
AK290561.1 |
- |
4, 5 |
379 |
|
AY950614.1 |
- |
4, 5 |
263 |
|
BC005319.1 |
- |
4, 5 |
320 |
3930700 |
BC013328.1 |
- |
4, 5 |
301 |
4134368 |
DA537533.1 |
- |
4, 5 |
379 |
|
DA539682.1 |
- |
4, 5 |
415 |
|
EU338231.1 |
- |
5, 6 |
383 |
|
EU338232.1 |
- |
4, 5 |
300 |
|
EU338233.1 |
- |
5, 6 |
383 |
|
EU338234.1 |
- |
3, 4 |
207 |
|
GU347002.1 |
- |
5, 6 |
407 |
|
GU347003.1 |
- |
5, 6 |
486 |
|
GU347004.1 |
- |
5, 6 |
464 |
|
GU347006.1 |
- |
4, 5 |
306 |
|
GU347008.1 |
- |
3, 4 |
201 |
|
L07251.1 |
- |
3, 4 |
167 |
|
M36281.1 |
- |
4, 5 |
273 |
|
U00177.1 |
- |
4, 5 |
263 |
|
U00178.1 |
- |
3, 4 |
167 |
|
U00179.1 |
- |
2, 3 |
68 |
|
X08054.1 |
- |
4, 5 |
319 |
|
X51798.1 |
- |
4, 5 |
287 |
|
View Full Product Details