Gene Transcripts
- Gene Symbol
- HERPUD1
- Entrez Gene ID
- 9709
- Gene Name
- homocysteine inducible ER protein with ubiquitin like domain 1
- Gene Aliases
- HERP, Mif1, SUP
- Chromosome Location
- Chr. 16: 56932090 - 56943881 on Build GRCh38
- UniGene ID
- Hs.146393
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001010989.2 |
NP_001010989.1
|
7, 8 |
1173 |
|
NM_001272103.1 |
NP_001259032.1
|
7, 8 |
1019 |
|
NM_014685.3 |
NP_055500.1
|
7, 8 |
1176 |
|
GenBank mRNA |
AB034989.1 |
- |
7, 8 |
1119 |
|
AF055001.1 |
- |
7, 8 |
1116 |
|
AF055003.1 |
- |
7, 8 |
916 |
|
AF217990.1 |
- |
4, 5 |
1457 |
|
AK223233.1 |
- |
7, 8 |
1119 |
|
AK291565.1 |
- |
7, 8 |
1119 |
|
AK292088.1 |
- |
7, 8 |
1119 |
|
AK302167.1 |
- |
6, 7 |
1080 |
|
BC000086.1 |
- |
7, 8 |
1111 |
3504515 |
BC008320.1 |
- |
7, 8 |
1110 |
3350209 |
BC009739.1 |
- |
5, 6 |
642 |
4095997 |
BC032673.1 |
- |
7, 8 |
1118 |
5575914 |
CR457116.1 |
- |
7, 8 |
1021 |
|
D14695.1 |
- |
7, 8 |
1114 |
|
DQ837586.1 |
- |
7, 8 |
961 |
|
View Full Product Details