Gene Transcripts

Gene Symbol
SFI1
Entrez Gene ID
9814
Gene Name
SFI1 centrin binding protein
Gene Aliases
PISD, PPP1R139, hSfi1p
Chromosome Location
Chr. 22: 31496139 - 31618554 on Build GRCh38
UniGene ID
Hs.62209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007467.2 NP_001007468.1 5 918
NM_001258325.1 NP_001245254.1 4 846
NM_001258326.1 NP_001245255.1 3 668
NM_001258327.1 NP_001245256.1 3 672
NM_014775.3 NP_055590.2 5 918
XM_005261868.2 XP_005261925.1 5 916
XM_006724390.2 XP_006724453.1 3 670
XM_011530570.2 XP_011528872.1 5 916
XM_011530571.2 XP_011528873.1 5 912
XM_011530574.2 XP_011528876.1 5 593
XM_011530575.2 XP_011528877.1 5 916
XM_011530576.2 XP_011528878.1 4 844
XM_011530577.2 XP_011528879.1 4 840
XM_011530579.2 XP_011528881.1 5 593
XM_011530580.2 XP_011528882.1 5 916
XM_011530581.2 XP_011528883.1 5 593
XM_011530582.2 XP_011528884.1 3 670
XM_011530583.2 XP_011528885.1 3 666
XM_017029139.1 XP_016884628.1 5 593
XM_017029140.1 XP_016884629.1 4 844
XM_017029141.1 XP_016884630.1 5 593
XM_017029142.1 XP_016884631.1 4 844
XM_017029143.1 XP_016884632.1 5 912
XM_017029144.1 XP_016884633.1 5 916
XM_017029145.1 XP_016884634.1 4 844
XM_017029146.1 XP_016884635.1 5 593
XM_017029147.1 XP_016884636.1 4 844
XM_017029148.1 XP_016884637.1 4 840
XM_017029149.1 XP_016884638.1 4 844
XM_017029150.1 XP_016884639.1 4 840
XM_017029151.1 XP_016884640.1 5 912
XM_017029152.1 XP_016884641.1 3 670
XM_017029153.1 XP_016884642.1 3 666
XM_017029154.1 XP_016884643.1 4 840
XM_017029155.1 XP_016884644.1 3 670
XM_017029156.1 XP_016884645.1 3 670
XM_017029158.1 XP_016884647.1 5 912
XM_017029159.1 XP_016884648.1 4 521
XM_017029161.1 XP_016884650.1 3 347
XM_017029162.1 XP_016884651.1 3 347
GenBank mRNA AB011114.2 - 5 782
AK292408.1 - 5 494
AK293987.1 - 4 459
AK303362.1 - 3 303
BC110814.1 - 3 269 6500257
BC129926.1 - 3 424 40124134
BC129945.1 - 4 598 40124130

View Full Product Details