Gene Transcripts

Gene Symbol
AP1M2
Entrez Gene ID
10053
Gene Name
adaptor related protein complex 1 mu 2 subunit
Gene Aliases
AP1-mu2, HSMU1B, MU-1B, MU1B, mu2
Chromosome Location
Chr. 19: 10572671 - 10587315 on Build GRCh38
UniGene ID
Hs.18894
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300887.1 NP_001287816.1 10, 11 1270
NM_005498.4 NP_005489.2 10, 11 1264
GenBank mRNA AF020797.2 - 10, 11 1227
AK023863.1 - 10, 11 1265
AK222946.1 - 10, 11 1230
AK315689.1 - 10, 11 1255
AW370781.1 - 2, 3 403
BC003387.2 - 10, 11 1202 3446133
BC003612.2 - 10, 11 1201 2988458
BC005021.1 - 10, 11 1270 3139433

View Full Product Details