Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005879.2 | NP_005870.2 | 15 | 1414 | |
XM_011533264.1 | XP_011531566.1 | 15 | 1508 | ||
XM_017005526.1 | XP_016861015.1 | 12 | 1142 | ||
GenBank mRNA | AB451320.1 | - | 15 | 1308 | |
AB451449.1 | - | 15 | 1308 | ||
AK225873.1 | - | 12 | 1093 | ||
AK292172.1 | - | 15 | 1402 | ||
BC000310.2 | - | 15 | 1395 | 2821007 | |
BC019283.1 | - | 15 | 1395 | 2821007 | |
BQ446539.1 | - | 1 | 625 | ||
U77845.1 | - | 15 | 1410 |