Gene Transcripts
- Gene Symbol
- CHEK2
- Entrez Gene ID
- 11200
- Gene Name
- checkpoint kinase 2
- Gene Aliases
- CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1
- Chromosome Location
- Chr. 22: 28687743 - 28741866 on Build GRCh38
- UniGene ID
- Hs.291363
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005735.1 |
NP_001005735.1
|
6, 7 |
891 |
|
NM_001257387.1 |
NP_001244316.1
|
6, 7 |
876 |
|
NM_007194.3 |
NP_009125.1
|
5, 6 |
762 |
|
NM_145862.2 |
NP_665861.1
|
5, 6 |
762 |
|
XM_011529839.2 |
XP_011528141.1
|
6, 7 |
913 |
|
XM_011529840.2 |
XP_011528142.1
|
6, 7 |
915 |
|
XM_011529844.2 |
XP_011528146.1
|
6, 7 |
923 |
|
XM_011529845.2 |
XP_011528147.1
|
4, 5 |
469 |
|
XM_017028560.1 |
XP_016884049.1
|
6, 7 |
877 |
|
XM_017028561.1 |
XP_016884050.1
|
2, 3 |
121 |
|
GenBank mRNA |
AB040105.1 |
- |
4, 5 |
690 |
|
AF086904.1 |
- |
4, 5 |
690 |
|
AF096279.1 |
- |
4, 5 |
690 |
|
AF174135.1 |
- |
4, 5 |
690 |
|
AF217975.1 |
- |
6, 7 |
1475 |
|
AJ131197.1 |
- |
4, 5 |
690 |
|
AK289360.1 |
- |
5, 6 |
741 |
|
AK290754.1 |
- |
5, 6 |
764 |
|
AY551297.1 |
- |
5, 6 |
819 |
|
AY551298.1 |
- |
4, 5 |
690 |
|
AY551299.1 |
- |
4, 5 |
690 |
|
AY551300.1 |
- |
4, 5 |
690 |
|
AY551301.1 |
- |
4, 5 |
580 |
|
AY551303.1 |
- |
2, 3 |
417 |
|
AY551305.1 |
- |
5, 6 |
804 |
|
BC004207.2 |
- |
5, 6 |
717 |
3532866 |
CR456418.1 |
- |
6, 7 |
890 |
|
View Full Product Details