Gene Transcripts

Gene Symbol
TRAM1
Entrez Gene ID
23471
Gene Name
translocation associated membrane protein 1
Gene Aliases
PNAS8, TRAM, TRAMP
Chromosome Location
Chr. 8: 70573218 - 70608459 on Build GRCh38
UniGene ID
Hs.491988
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317804.1 NP_001304733.1 2, 3 296
NM_014294.5 NP_055109.1 1, 2 379
XM_017013266.1 XP_016868755.1 1, 2 373
GenBank mRNA AK292777.1 - 1, 2 163
AK303411.1 - 2, 3 224
BC000687.2 - 1, 2 201 3347823
BC020283.1 - 1, 2 194
BC037738.1 - 1, 2 291 5261490
BI465346.1 - 1, 2 365
BT007359.1 - 1, 2 119
CR541654.1 - 1, 2 119
CR541739.1 - 1, 2 119
X63679.1 - 1, 2 240

View Full Product Details