Gene Transcripts

Gene Symbol
RAB30
Entrez Gene ID
27314
Gene Name
RAB30, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 11: 82973133 - 83071923 on Build GRCh38
UniGene ID
Hs.40758
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286059.1 NP_001272988.1 5 823
NM_001286060.1 NP_001272989.1 5 658
NM_001286061.1 NP_001272990.1 5 556
NM_014488.4 NP_055303.2 6 759
GenBank mRNA AF498956.1 - 4 417
AK291376.1 - 5 818
AK312323.1 - 5 564
BC014213.2 - 6 678 4763140
BP336371.1 - 5 555
BX641138.1 - 5 603
CR542106.1 - 4 417
U57092.1 - 4 435

View Full Product Details