Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256007.2 | NP_001242936.1 | 3 | 1121 | |
NM_001256008.2 | NP_001242937.1 | 2 | 1075 | ||
NM_001256009.2 | NP_001242938.1 | 2 | 1075 | ||
NM_001256010.2 | NP_001242939.1 | 2 | 948 | ||
NM_001256011.2 | NP_001242940.1 | 3 | 809 | ||
NM_015723.4 | NP_056538.1 | 4 | 1233 | ||
XM_005250396.4 | XP_005250453.1 | 3 | 2077 | ||
XM_011516274.2 | XP_011514576.1 | 3 | 1950 | ||
XM_011516275.2 | XP_011514577.1 | 3 | 1950 | ||
XM_017012261.1 | XP_016867750.1 | 2 | 1904 | ||
GenBank mRNA | AB041261.1 | - | 2 | 901 | |
AF263613.1 | - | 3 | 927 | ||
AK024335.1 | - | 3 | 809 | ||
AK291755.1 | - | 3 | 938 | ||
AL834147.1 | - | 2 | 828 | ||
BC032999.2 | - | 4 | 1109 | 4829433 | |
BX647865.1 | - | 2 | 794 |