Gene Transcripts

Gene Symbol
ZDHHC9
Entrez Gene ID
51114
Gene Name
zinc finger DHHC-type containing 9
Gene Aliases
CGI89, CXorf11, DHHC9, MMSA1, MRXSZ, ZDHHC10, ZNF379, ZNF380
Chromosome Location
Chr. X: 129803288 - 129843934 on Build GRCh38
UniGene ID
Hs.193566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008222.2 NP_001008223.1 4 871
NM_016032.3 NP_057116.2 5 851
XM_011531348.2 XP_011529650.1 5 838
GenBank mRNA AB209807.1 - 4 790
AF151847.1 - 5 739
AK001112.1 - 5 801
AK001424.1 - 2 185
AK001524.1 - 4 602
AK096967.1 - 5 502
AL161962.1 - 5 728
AY358558.1 - 5 783
AY952881.8 - 5 823
BC000035.2 - 4 745 3506601
BC003128.1 - 5 789 2957911
BC006200.2 - 5 785 2964425
BC012826.2 - 5 785 2964425

View Full Product Details