Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005739.1 | NP_001005739.1 | 21 | 2784 | |
NM_016516.2 | NP_057600.2 | 21 | 2820 | ||
XM_006712029.3 | XP_006712092.1 | 20 | 2680 | ||
XM_017004316.1 | XP_016859805.1 | 20 | 2500 | ||
GenBank mRNA | AF102177.1 | - | 20 | 2677 | |
AK002205.1 | - | 9 | 925 | ||
AK025545.1 | - | 15 | 1712 | ||
AK094019.1 | - | 6 | 1231 | ||
AK124436.1 | - | 10 | 3968 | ||
AK292669.1 | - | 22 | 2828 | ||
AK292889.1 | - | 22 | 2879 | ||
AL359939.1 | - | 8 | 642 | ||
AY444798.1 | - | 20 | 2677 | ||
BC030275.1 | - | 21 | 2787 | 3917367 | |
BC041868.1 | - | 18 | 2788 | 5271464 | |
CR749701.1 | - | 8 | 642 |