Gene Transcripts

Gene Symbol
SIDT1
Entrez Gene ID
54847
Gene Name
SID1 transmembrane family member 1
Gene Aliases
SID-1, SID1
Chromosome Location
Chr. 3: 113531774 - 113629579 on Build GRCh38
UniGene ID
Hs.591291
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308350.1 NP_001295279.1 8 1506
NM_001322294.1 NP_001309223.1 8 1506
NM_001322295.1 NP_001309224.1 7 1422
NM_001322296.1 NP_001309225.1 6 1320
NM_001322297.1 NP_001309226.1 7 1384
NM_001322298.1 NP_001309227.1 7 1483
NM_001322299.1 NP_001309228.1 8 1525
NM_001322300.1 NP_001309229.1 6 1320
NM_017699.2 NP_060169.2 8 1506
NR_136280.1 9 1586
NR_136281.1 7 1483
NR_136282.1 8 1506
NR_136283.1 8 1506
NR_136284.1 6 1320
NR_136285.1 6 1320
XM_011512939.2 XP_011511241.1 8 1466
XM_011512941.2 XP_011511243.1 8 1466
XM_017006664.1 XP_016862153.1 6 1397
XM_017006665.1 XP_016862154.1 6 1897
XM_017006666.1 XP_016862155.1 5 374
XM_017006667.1 XP_016862156.1 3 241
GenBank mRNA BC117222.1 - 8 894
BC143776.1 - 8 894
BC143778.1 - 8 913

View Full Product Details